Assay Details
| Gene Symbol: mrt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067120.5 | NP_499521.1 | 4 - 5 | 599 | 67 | |
| GenBank mRNA | AF076843.1 | - | 3 - 4 | 409 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrt-2| Gene Symbol: mrt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067120.5 | NP_499521.1 | 4 - 5 | 599 | 67 | |
| GenBank mRNA | AF076843.1 | - | 3 - 4 | 409 | 67 | |
Set Membership: |
|