Assay Details
| Gene Symbol: mrt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067120.5 | NP_499521.1 | 7 - 8 | 894 | 98 | |
| GenBank mRNA | AF076843.1 | - | 6 - 7 | 704 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrt-2| Gene Symbol: mrt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067120.5 | NP_499521.1 | 7 - 8 | 894 | 98 | |
| GenBank mRNA | AF076843.1 | - | 6 - 7 | 704 | 98 | |
Set Membership: |
|