Assay Details
| Gene Symbol: gbf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379938.1 | NP_001366879.1 | 16 - 17 | 5200 | 64 | |
| NM_001379939.1 | NP_001368124.1 | 2 - 3 | 710 | 64 | ||
More Information
Set Membership: |
|

gbf-1
Genomic Map