Assay Details
Gene Symbol: ntl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083183.3 | NP_001076652.1 | 2 - 3 | 196 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
