Assay Details
Gene Symbol: ntl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083183.3 | NP_001076652.1 | 5 - 6 | 1365 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
