Assay Details
Gene Symbol: taf-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268215.3 | NP_001255144.1 | 1 - 2 | 105 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
