Assay Details
Gene Symbol: taf-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268215.3 | NP_001255144.1 | 5 - 6 | 703 | 75 | |
NM_001268216.3 | NP_001255145.1 | 4 - 5 | 600 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
