Assay Details
| Gene Symbol: faah-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330969.2 | NP_001317730.1 | 6 - 7 | 1540 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

faah-5| Gene Symbol: faah-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330969.2 | NP_001317730.1 | 6 - 7 | 1540 | 77 | |
Set Membership: |
|