Assay Details
Gene Symbol: top-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067157.6 | NP_499558.1 | 6 - 7 | 1202 | 71 | |
GenBank mRNA | AF057032.1 | - | 6 - 7 | 1202 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
