Assay Details
Gene Symbol: top-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067157.6 | NP_499558.1 | 8 - 9 | 2031 | 65 | |
GenBank mRNA | AF057032.1 | - | 8 - 9 | 2031 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
