Assay Details
| Gene Symbol: ung-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268220.3 | NP_001255149.1 | 1 - 2 | 60 | 83 | |
| NM_067160.7 | NP_499561.1 | 2 - 3 | 199 | 83 | ||
More Information
Set Membership: |
|

ung-1
Genomic Map