Assay Details
| Gene Symbol: brp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027780.7 | NP_001022951.1 | 2 - 3 | 260 | 62 | |
| NM_001268223.3 | NP_001255152.1 | 1 - 2 | 149 | 62 | ||
More Information
Set Membership: |
|

brp-1
Genomic Map