Assay Details
| Gene Symbol: brp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027780.7 | NP_001022951.1 | 5 - 6 | 1158 | 63 | |
| NM_001268223.3 | NP_001255152.1 | 4 - 5 | 1047 | 63 | ||
More Information
Set Membership: |
|

brp-1
Genomic Map