Assay Details
Gene Symbol: brp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027780.7 | NP_001022951.1 | 6 - 7 | 1351 | 119 | |
NM_001268223.3 | NP_001255152.1 | 5 - 6 | 1240 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
