Assay Details
| Gene Symbol: tim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067193.7 | NP_499594.2 | 3 - 4 | 393 | 94 | |
| GenBank mRNA | AF183401.1 | - | 2 - 3 | 251 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tim-1| Gene Symbol: tim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067193.7 | NP_499594.2 | 3 - 4 | 393 | 94 | |
| GenBank mRNA | AF183401.1 | - | 2 - 3 | 251 | 94 | |
Set Membership: |
|