Assay Details
Gene Symbol: mrp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067197.4 | NP_499598.2 | 18 - 19 | 3844 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
