Assay Details
| Gene Symbol: pph-4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067202.6 | NP_499603.1 | 2 - 3 | 233 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pph-4.1| Gene Symbol: pph-4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067202.6 | NP_499603.1 | 2 - 3 | 233 | 59 | |
Set Membership: |
|