Assay Details
| Gene Symbol: tat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027723.3 | NP_001022894.1 | 14 - 15 | 3151 | 66 | |
| NM_001268236.3 | NP_001255165.1 | 14 - 15 | 3003 | 66 | ||
| NM_001379952.1 | NP_001367088.1 | 13 - 14 | 2848 | 66 | ||
More Information
Set Membership: |
|

tat-1
Genomic Map