Assay Details
Gene Symbol: tat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027723.3 | NP_001022894.1 | 16 - 17 | 3496 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
