Assay Details
Gene Symbol: tat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027723.3 | NP_001022894.1 | 17 - 18 | 3630 | 70 | |
NM_001268236.3 | NP_001255165.1 | 18 - 19 | 4150 | 70 | ||
NM_001379952.1 | NP_001367088.1 | 18 - 19 | 4068 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
