Assay Details
Gene Symbol: tat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027723.3 | NP_001022894.1 | 7 - 8 | 1232 | 94 | |
NM_001268236.3 | NP_001255165.1 | 7 - 8 | 1084 | 94 | ||
NM_001379952.1 | NP_001367088.1 | 6 - 7 | 929 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
