Assay Details
| Gene Symbol: tat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027723.3 | NP_001022894.1 | 8 - 9 | 1355 | 93 | |
| NM_001268236.3 | NP_001255165.1 | 8 - 9 | 1207 | 93 | ||
| NM_001379952.1 | NP_001367088.1 | 7 - 8 | 1052 | 93 | ||
More Information
Set Membership: |
|

tat-1
Genomic Map