Assay Details
| Gene Symbol: pie-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268237.2 | NP_001255166.1 | 4 - 5 | 612 | 82 | |
| GenBank mRNA | U62896.1 | - | 4 - 5 | 621 | 82 | |
More Information
Set Membership: |
|

pie-1
Genomic Map