Assay Details
| Gene Symbol: bre-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268259.3 | NP_001255188.1 | 6 - 7 | 839 | 105 | |
| NM_001268260.1 | NP_001255189.1 | 6 - 7 | 827 | 105 | ||
| NM_001379974.2 | NP_001367175.1 | 4 - 5 | 704 | 105 | ||
| NM_067314.4 | NP_499715.2 | 6 - 7 | 839 | 105 | ||
| GenBank mRNA | AY533304.1 | - | 4 - 5 | 704 | 105 | |
More Information
Set Membership: |
|

bre-2
Genomic Map