Assay Details
| Gene Symbol: cdk-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067382.8 | NP_499783.1 | 2 - 3 | 214 | 60 | |
| GenBank mRNA | AF129111.1 | - | 2 - 3 | 119 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdk-5| Gene Symbol: cdk-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067382.8 | NP_499783.1 | 2 - 3 | 214 | 60 | |
| GenBank mRNA | AF129111.1 | - | 2 - 3 | 119 | 60 | |
Set Membership: |
|