Assay Details
| Gene Symbol: mdt-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067446.7 | NP_499847.1 | 10 - 11 | 1595 | 60 | |
| Gene Symbol: pot-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379991.1 | NP_001368150.1 | 3 - 4 | 382 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379991.1 transcripts.
Set Membership: |
|

mdt-29
Genomic Map