Assay Details
| Gene Symbol: mdt-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067446.7 | NP_499847.1 | 11 - 12 | 1829 | 63 | |
| Gene Symbol: pot-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379991.1 | NP_001368150.1 | 4 - 5 | 616 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379991.1 transcripts.
Set Membership: |
|

mdt-29
Genomic Map