Assay Details
| Gene Symbol: spl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067512.8 | NP_499913.1 | 6 - 7 | 900 | 76 | |
| GenBank mRNA | AF144639.1 | - | 6 - 7 | 894 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spl-1| Gene Symbol: spl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067512.8 | NP_499913.1 | 6 - 7 | 900 | 76 | |
| GenBank mRNA | AF144639.1 | - | 6 - 7 | 894 | 76 | |
Set Membership: |
|