Assay Details
| Gene Symbol: ssna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028191.8 | NP_001023362.1 | 2 - 3 | 923 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ssna-1| Gene Symbol: ssna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028191.8 | NP_001023362.1 | 2 - 3 | 923 | 72 | |
Set Membership: |
|