Assay Details
| Gene Symbol: plk-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068795.7 | NP_501196.1 | 1 - 2 | 312 | 110 | |
| GenBank mRNA | AF059024.1 | - | 1 - 2 | 312 | 110 | |
More Information
Set Membership: |
|

plk-3
Genomic Map