Assay Details
| Gene Symbol: rod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068799.7 | NP_501200.1 | 3 - 4 | 278 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rod-1| Gene Symbol: rod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068799.7 | NP_501200.1 | 3 - 4 | 278 | 153 | |
Set Membership: |
|