Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307219.4 | NP_001294148.2 | 12 - 13 | 16330 | 138 | |
| NM_001307220.4 | NP_001294149.2 | 12 - 13 | 16330 | 138 | ||
More Information
Set Membership: |
|

cla-1
Genomic Map