Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307214.3 | NP_001294143.1 | 2 - 3 | 141 | 80 | |
| NM_001307215.3 | NP_001294144.1 | 2 - 3 | 141 | 80 | ||
| NM_001307219.4 | NP_001294148.2 | 15 - 16 | 18981 | 80 | ||
| NM_001307220.4 | NP_001294149.2 | 15 - 16 | 18981 | 80 | ||
More Information
Set Membership: |
|

cla-1
Genomic Map