Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307215.3 | NP_001294144.1 | 21 - 22 | 7133 | 132 | |
| NM_001307219.4 | NP_001294148.2 | 34 - 35 | 25973 | 132 | ||
| NM_001383125.1 | 14 - 15 | 6463 | 132 | |||
More Information
Set Membership: |
|

cla-1
Genomic Map