Assay Details
| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307215.3 | NP_001294144.1 | 22 - 23 | 7442 | 86 | |
| NM_001307219.4 | NP_001294148.2 | 35 - 36 | 26282 | 86 | ||
| NM_001383125.1 | 15 - 16 | 6772 | 86 | |||
More Information
Set Membership: |
|

cla-1
Genomic Map