Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 18 - 19 | 5070 | 67 | |
| NM_001028000.5 | NP_001023171.1 | 16 - 17 | 4902 | 67 | ||
| NM_001028001.5 | NP_001023172.1 | 18 - 19 | 5027 | 67 | ||
| NM_001028002.4 | NP_001023173.1 | 17 - 18 | 4998 | 67 | ||
| NM_001268376.4 | NP_001255305.1 | 17 - 18 | 4932 | 67 | ||
| NM_001380339.1 | NP_001367466.1 | 17 - 18 | 4932 | 67 | ||
| GenBank mRNA | AF168688.1 | - | 17 - 18 | 4998 | 67 | |
| AJ243179.2 | - | 17 - 18 | 5020 | 67 | ||
| AJ243180.2 | - | 18 - 19 | 5093 | 67 | ||
| AJ243181.2 | - | 17 - 18 | 5013 | 67 | ||
| AJ243182.2 | - | 18 - 19 | 5047 | 67 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map