Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 22 - 23 | 5664 | 116 | |
| NM_001028000.5 | NP_001023171.1 | 20 - 21 | 5496 | 116 | ||
| NM_001028001.5 | NP_001023172.1 | 23 - 24 | 5654 | 116 | ||
| NM_001028002.4 | NP_001023173.1 | 21 - 22 | 5592 | 116 | ||
| NM_001268376.4 | NP_001255305.1 | 22 - 23 | 5589 | 116 | ||
| GenBank mRNA | AF168688.1 | - | 21 - 22 | 5592 | 116 | |
| AJ243179.2 | - | 21 - 22 | 5614 | 116 | ||
| AJ243180.2 | - | 22 - 23 | 5687 | 116 | ||
| AJ243181.2 | - | 21 - 22 | 5607 | 116 | ||
| AJ243182.2 | - | 23 - 24 | 5674 | 116 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map