Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 32 - 33 | 8597 | 147 | |
| NM_001028000.5 | NP_001023171.1 | 30 - 31 | 8429 | 147 | ||
| NM_001028001.5 | NP_001023172.1 | 32 - 33 | 8560 | 147 | ||
| NM_001028002.4 | NP_001023173.1 | 31 - 32 | 8525 | 147 | ||
| NM_001268376.4 | NP_001255305.1 | 32 - 33 | 8522 | 147 | ||
| GenBank mRNA | AF168688.1 | - | 31 - 32 | 8525 | 147 | |
| AJ243179.2 | - | 31 - 32 | 8547 | 147 | ||
| AJ243180.2 | - | 32 - 33 | 8620 | 147 | ||
| AJ243181.2 | - | 31 - 32 | 8540 | 147 | ||
| AJ243182.2 | - | 32 - 33 | 8580 | 147 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map