Assay Details
| Gene Symbol: itr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027999.4 | NP_001023170.1 | 4 - 5 | 538 | 149 | |
| NM_001028000.5 | NP_001023171.1 | 2 - 3 | 370 | 149 | ||
| NM_001028001.5 | NP_001023172.1 | 3 - 4 | 468 | 149 | ||
| NM_001028002.4 | NP_001023173.1 | 3 - 4 | 466 | 149 | ||
| NM_001268376.4 | NP_001255305.1 | 3 - 4 | 400 | 149 | ||
| NM_001380339.1 | NP_001367466.1 | 3 - 4 | 400 | 149 | ||
| GenBank mRNA | AF168688.1 | - | 3 - 4 | 466 | 149 | |
| AJ243179.2 | - | 3 - 4 | 488 | 149 | ||
| AJ243180.2 | - | 4 - 5 | 561 | 149 | ||
| AJ243181.2 | - | 3 - 4 | 481 | 149 | ||
| AJ243182.2 | - | 3 - 4 | 488 | 149 | ||
More Information
Set Membership: |
|

itr-1
Genomic Map