Assay Details
Gene Symbol: imp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307234.3 | NP_001294163.1 | 3 - 4 | 454 | 77 | |
NM_001372874.1 | NP_001360383.1 | 6 - 7 | 1052 | 77 | ||
NM_068885.8 | NP_501286.2 | 6 - 7 | 901 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
