Assay Details
| Gene Symbol: imp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307234.3 | NP_001294163.1 | 4 - 5 | 549 | 116 | |
| NM_001372874.1 | NP_001360383.1 | 7 - 8 | 1147 | 116 | ||
| NM_068885.8 | NP_501286.2 | 7 - 8 | 996 | 116 | ||
More Information
Set Membership: |
|

imp-3
Genomic Map