Assay Details
| Gene Symbol: imp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307234.3 | NP_001294163.1 | 6 - 7 | 1017 | 87 | |
| NM_001307235.3 | NP_001294164.1 | 1 - 2 | 195 | 87 | ||
| NM_001372874.1 | NP_001360383.1 | 9 - 10 | 1615 | 87 | ||
| NM_068885.8 | NP_501286.2 | 9 - 10 | 1464 | 87 | ||
More Information
Set Membership: |
|

imp-3
Genomic Map