Assay Details
| Gene Symbol: sgo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307239.2 | NP_001294168.1 | 2 - 3 | 240 | 86 | |
| NM_068889.5 | NP_501290.2 | 1 - 2 | 183 | 86 | ||
More Information
Set Membership: |
|

sgo-1
Genomic Map