Assay Details
Gene Symbol: sgo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307239.2 | NP_001294168.1 | 4 - 5 | 377 | 148 | |
NM_068889.5 | NP_501290.2 | 3 - 4 | 320 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
