Assay Details
Gene Symbol: sgo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307239.2 | NP_001294168.1 | 6 - 7 | 772 | 82 | |
NM_068889.5 | NP_501290.2 | 5 - 6 | 715 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
