Assay Details
| Gene Symbol: gcy-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068923.7 | NP_501324.2 | 1 - 2 | 389 | 74 | |
| GenBank mRNA | AB201388.1 | - | 1 - 2 | 318 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-8| Gene Symbol: gcy-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068923.7 | NP_501324.2 | 1 - 2 | 389 | 74 | |
| GenBank mRNA | AB201388.1 | - | 1 - 2 | 318 | 74 | |
Set Membership: |
|