Assay Details
| Gene Symbol: gcy-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068923.7 | NP_501324.2 | 6 - 7 | 1539 | 104 | |
| GenBank mRNA | AB201388.1 | - | 6 - 7 | 1468 | 104 | |
More Information
Set Membership: |
|

gcy-8
Genomic Map