Assay Details
Gene Symbol: pct-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026141.6 | NP_001021312.1 | 7 - 8 | 1311 | 66 | |
NM_068971.7 | NP_501372.1 | 6 - 7 | 631 | 66 | ||
NR_131581.1 | 6 - 7 | 1244 | 66 | |||
GenBank mRNA | AF129110.1 | - | 5 - 6 | 577 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
