Assay Details
Gene Symbol: pct-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026141.6 | NP_001021312.1 | 9 - 10 | 1690 | 163 | |
NM_068971.7 | NP_501372.1 | 8 - 9 | 1010 | 163 | ||
NR_131581.1 | 8 - 9 | 1623 | 163 | |||
GenBank mRNA | AF129110.1 | - | 7 - 8 | 956 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
