Assay Details
| Gene Symbol: svh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068978.6 | NP_501379.2 | 15 - 16 | 2680 | 79 | |
| GenBank mRNA | AB693146.1 | - | 15 - 16 | 2680 | 79 | |
More Information
Set Membership: |
|

svh-1
Genomic Map